Learn More
R&D Systems™ Human Endostatin Quantikine ELISA Kit, R&D Systems™
Human Endostatin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich 0.31 - 10ng/mL
Assay-Empfindlichkeit: 0.063ng/mL
Probentyp Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
The Human Endostatin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Endostatin in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (50 ul).
Spezifikation
Spezifikation
0.31 - 10ng/mL
0.063ng/mL
Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum