Learn More
Abnova™ Human ETS2 Full-length ORF (NP_005230.1, 1 a.a. - 469 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002114-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes (Dwyer et al., 2007 [PubMed 17986575]).[supplied by OMIM]
Sequence: MNDFGIKNMDQVAPVANSYRGTLKRQPAFDTFDGSLFAVFPSLNEEQTLQEVPTGLDSISHDSANCELPLLTPCSKAVMSQALKATFSGFKKEQRRLGIPKNPWLWSEQQVCQWLLWATNEFSLVNVNLQRFGMNGQMLCNLGKERFLELAPDFVGDILWEHLEQMIKENQEKTEDQYEENSHLTSVPHWINSNTLGFGTEQAPYGMQTQNYPKGGLLDSMCPASTPSVLSSEQEFQMFPKSRLSSVSVTYCSVSQDFPGSNLNLLTNNSGTPKDHDSPENGADSFESSDSLLQSWNSQSSLLDVQRVPSFESFEDDCSQSLCLNKPTMSFKDYIQERSDPVEQGKPVIPAAVLAGFTGSGPIQLWQFLLELLSDKSCQSFISWTGDGWEFKLADPDEVARRWGKRKNKPKMNYEKLSRGLRYYYDKNIIHKTSGKRYVYRFVCDLQNLLGFTPEELHAILGVQPDTEDSpezifikation
NP_005230.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ETS2IT1 | |
ETS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2114 | |
ETS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNDFGIKNMDQVAPVANSYRGTLKRQPAFDTFDGSLFAVFPSLNEEQTLQEVPTGLDSISHDSANCELPLLTPCSKAVMSQALKATFSGFKKEQRRLGIPKNPWLWSEQQVCQWLLWATNEFSLVNVNLQRFGMNGQMLCNLGKERFLELAPDFVGDILWEHLEQMIKENQEKTEDQYEENSHLTSVPHWINSNTLGFGTEQAPYGMQTQNYPKGGLLDSMCPASTPSVLSSEQEFQMFPKSRLSSVSVTYCSVSQDFPGSNLNLLTNNSGTPKDHDSPENGADSFESSDSLLQSWNSQSSLLDVQRVPSFESFEDDCSQSLCLNKPTMSFKDYIQERSDPVEQGKPVIPAAVLAGFTGSGPIQLWQFLLELLSDKSCQSFISWTGDGWEFKLADPDEVARRWGKRKNKPKMNYEKLSRGLRYYYDKNIIHKTSGKRYVYRFVCDLQNLLGFTPEELHAILGVQPDTED | |
RUO | |
ETS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |