missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FAM109B Full-length ORF (AAI04177.1, 1 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00150368-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MKLNERSVAHYALSDSPADHMGFLRTWGGPGTPPTPSGTGRRCWFVLKGNLLFSFESREGRAPLSLVVLEGCTVELAEAPVPEEFAFAICFDAPGVRPHLLAAEGPAAQEAWVKVLSRASFGYMRLVVRELESQLQDARQSLALQRRSSWKSVASRCKPQAPNHRAAGLENGHCLSKDSSPVGLVEEGGSRSAGWGLAEWELQGPASLLLGKGQSPVSPETSCFSTLHDWYGQEIVELRQCWQKRAQGSHSKCEEQDRPSpezifikation
AAI04177.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686J07229/MGC125831/MGC125832 | |
FAM109B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
150368 | |
FAM109B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKLNERSVAHYALSDSPADHMGFLRTWGGPGTPPTPSGTGRRCWFVLKGNLLFSFESREGRAPLSLVVLEGCTVELAEAPVPEEFAFAICFDAPGVRPHLLAAEGPAAQEAWVKVLSRASFGYMRLVVRELESQLQDARQSLALQRRSSWKSVASRCKPQAPNHRAAGLENGHCLSKDSSPVGLVEEGGSRSAGWGLAEWELQGPASLLLGKGQSPVSPETSCFSTLHDWYGQEIVELRQCWQKRAQGSHSKCEEQDRP | |
RUO | |
FAM109B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |