Learn More
Invitrogen™ Human FAM132A (aa 70-138) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54980 (PA5-54980. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Zugriffsnummer | Q5T7M4 |
| Konzentration | ≥5.0 mg/mL |
| Zur Verwendung mit (Anwendung) | Blocking Assay, Control |
| Zusammensetzung | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-ID (Entrez) | 388581 |
| Name | Human FAM132A (aa 70-138) Control Fragment |
| Menge | 100 μl |
| Kennzeichnung | RUO |
| Gen-Alias | Adipolin; Adipolin cleaved form; Adipolin fC1QTNF12; Adipolin fCTRP12; Adipolin full-length form; Adipolin gC1QTNF12; Adipolin gCTRP12; Adipose-derived insulin-sensitizing factor; C1q and TNF related 12; C1q and TNF related protein 12; C1q domain containing 2; C1q/TNF-related protein 12; C1QDC2; C1QTNF12; Complement C1q tumor necrosis factor-related protein 12; CTRP12; FAM132A; family with sequence similarity 132 member A; family with sequence similarity 132, member A; protein FAM132A; zgc:158358 |
| Gebräuchliche Bezeichnung | FAM132A |
| Show More |