missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FARSLA Partial ORF (NP_004452, 101 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00002193-Q01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16167974

  • 335.00 € / 10 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. This gene encodes a product which is similar to the catalytic subunit of prokaryotic and Saccharomyces cerevisiae phenylalanyl-tRNA synthetases (PheRS). This gene product has been shown to be expressed in a tumor-selective and cell cycle stage- and differentiation-dependent manner, the first member of the tRNA synthetase gene family shown to exhibit this type of regulated expression [provided by RefSeq]

Sequence: KVGFSKAMSNKWIRVDKSAADGPRVFRVVDSMEDEVQRRLQLVRGGQAEKLGEKERSELRKRKLLAEVTLKTYWVSKGSAFSTSISKQETELSPEMISSGS
Spezifikation

Spezifikation

NP_004452
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KVGFSKAMSNKWIRVDKSAADGPRVFRVVDSMEDEVQRRLQLVRGGQAEKLGEKERSELRKRKLLAEVTLKTYWVSKGSAFSTSISKQETELSPEMISSGS
RUO
FARSA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2193
FARSLA (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CML33/FARSL/FARSLA/FRSA/PheHA
FARSA
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote