Learn More
Abnova™ Human FCAR Partial ORF (AAH27953, 24 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002204-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: GDFPMPFISAKSSPVIPLDGSVKIQCQAIREAYLTQLMIIKNSTYREIGRRLKFWNETDPEFVIDHMDANKAGRYQCQYRIGHYRFRYSDTLELVVTGLYGKPFLSADRGSpezifikation
AAH27953 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GDFPMPFISAKSSPVIPLDGSVKIQCQAIREAYLTQLMIIKNSTYREIGRRLKFWNETDPEFVIDHMDANKAGRYQCQYRIGHYRFRYSDTLELVVTGLYGKPFLSADRG | |
RUO | |
FCAR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2204 | |
FCAR (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD89 | |
FCAR | |
Recombinant | |
wheat germ expression system |