Learn More
Abnova™ Human FCRLM2 Partial ORF (-, 89 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00127943-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
FCRL2 belongs to the Fc receptor family. Fc receptors are involved in phagocytosis, antibody-dependent cell cytotoxicity, immediate hypersensitivity, and transcytosis of immunoglobulins via their ability to bind immunoglobulin (Ig) constant regions (Chikaev et al., 2005 [PubMed 15676285]).[supplied by OMIM]
Sequence: DSLASCKAGAASPILGCRTRAECQSGCDMKRLAWSLFLSSFPRPSWTRSPCPTFQKAPLPPPRGHTASSPPPLVCGSARSPRGKQESpezifikation
NP_689591.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.2kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DSLASCKAGAASPILGCRTRAECQSGCDMKRLAWSLFLSSFPRPSWTRSPCPTFQKAPLPPPRGHTASSPPPLVCGSARSPRGKQE | |
RUO | |
FCRLB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
127943 | |
FCRLM2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FCRL2/FCRLM2/FCRLY/FLJ31052/FREB-2/FcRY/RP11-474I16.6 | |
FCRLB | |
Recombinant | |
wheat germ expression system |