Learn More
ACROBiosystems Human Fetuin B Protein, His Tag
Marke: ACROBiosystems FEB-H52H7-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Fetuin-B (FETUB), a second member of the fetuin family in mammals, is also known as Fetuin-like protein IRL685, 16G2 and Gugu, which is expressed in liver and testis and contains 2 cystatin fetuin-B-type domains. FETUB is a protease inhibitor required for egg fertilization. FETUB is also required to prevent premature zona pellucida hardening before fertilization, probably by inhibiting the protease activity of ASTL, a protease that mediates the cleavage of ZP2 and triggers zona pellucida hardening (By similarity). As a liver-derived plasma protein, FETUB is essential for fertilization.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.4 kDa. The protein migrates as 53-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH69670 | |
100 μg | |
Cys 16 - Pro 382 | |
FETUB,Fetuin-B,16G2,Gugu | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
42.4 kDa | |
Human Fetuin B, His Tag (FEB-H52H7) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Pro 382 (Accession # AAH69670). | |
-20°C | |
Fetuin B | |
Human | |
50 mM Tris, 150 mM NaCl, pH8.0 | |
HEK293 | |
Other Recombinant Proteins |