missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human FGF basic Quantikine ELISA Kit, R&D Systems™

A 4.5 hour solid phase ELISA designed to measure human FGF basic in cell culture supernates, serum, and plasma. It contains E. coli-expressed recombinant human FGF basic and antibodies raised against the recombinant factor.

Marke:  R&D Systems™ SFB50

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412776

  • 4348.06 € / 6 Stück


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
Sensitivity: 3pg/mL
Specificity: Natural and recombinant human FGF basic
Cross Reactivity: Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules.

Spezifikation

Spezifikation

10.00 - 640pg/mL
Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules
2247
6 Plates
3pg/mL
Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (100uL), Serum (100uL), EDTA Plasma (100uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Videos