missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXD4L1 Partial ORF (NP_036316.1, 285 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00200350-Q01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16162377

  • 335.00 € / 10 Mikrogramm
Verfügbar ab: 27-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene is a member of the forkhead/winged-helix (FOX) family of transcription factors with highly conserved FOX DNA-binding domains. Members of the FOX family of transcription factors are regulators of embryogenesis and may play a role in human cancer. This gene lies in a region of chromosome 2 that surrounds the site where two ancestral chromosomes fused to form human chromosome 2. This region is duplicated elsewhere in the human genome, primarily in subtelomeric and pericentromeric locations, thus mutiple copies of this gene have been found. [provided by RefSeq]

Sequence: EGADLATPGTLPVLQPSLGPQPWEEGKGLASPPGGGCISFSIESIMQGVRGAGTGAAQSLSPTAWSYCPLLQRPSSLSDNFAATAAASGGGLRQRLRSHQ
Spezifikation

Spezifikation

NP_036316.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EGADLATPGTLPVLQPSLGPQPWEEGKGLASPPGGGCISFSIESIMQGVRGAGTGAAQSLSPTAWSYCPLLQRPSSLSDNFAATAAASGGGLRQRLRSHQ
RUO
FOXD4L1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
200350
FOXD4L1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FOXD5/bA395L14.1
FOXD4L1
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote