missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FOXM1 Partial ORF (NP_973731.1, 702 a.a. - 801 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002305-Q01.10ug
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Sequence: LQSAPPLESPQRLLSSEPLDLISVPFGNSSPSDIDVPKPGSPEPQVSGLAANRSLTEGLVLDTMNDSLSKILLDISFPGLDEDPLGPDNINWSQFIPELQSpezifikation
NP_973731.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LQSAPPLESPQRLLSSEPLDLISVPFGNSSPSDIDVPKPGSPEPQVSGLAANRSLTEGLVLDTMNDSLSKILLDISFPGLDEDPLGPDNINWSQFIPELQ | |
RUO | |
FOXM1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
2305 | |
FOXM1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKHL16|FOXM1B|HFH-11|HFH11|HNF-3|INS-1|MPHOSPH2|MPP-2|MPP2|PIG29|TGT3|TRIDENT | |
FOXM1 | |
Yes |