Learn More
Abnova™ Human GABRA5 Full-length ORF (-, 1 a.a. - 462 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002558-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Transcript variants utilizing three different alternative non-coding first exons have been described. [provided by RefSeq]
Sequence: MDNGMFSGFIMIKNLLLFCISMNLSSHFGFSQMPTSSVKDETNDNITIFTRILDGLLDGYDNRLRPGLGERITQVRTDIYVTSFGPVSDTEMEYTIDVFFRQSWKDERLRFKGPMQRLPLNNLLASKIWTPDTFFHNGKKSIAHNMTTPNKLPRLEDDGTLLYTMRLTISAECPMQLEDFPMDAHACPLKFGSYAYPNSEVVYVWTNGSTKSVVVAEDGSRLNQYHLMGQTVGTENISTSTGEYTIMTAHFHLKRKIGYFVIQTYLPCIMTVILSQVSFWLNRESVPARTVFGVTTVLTMTTLSISARNSLPKVAYATAMDWFIAVCYAFVFSALIEFATVNYFTKRGWAWDGKKALEAAKIKKKREVILNKSTNAFTTGKMSHPPNIPKEQTPAGTSNTTSVSVKPSEEKTSESKKTYNSISKIDKMSRIVFPVLFGTFNLVYWATYLNREPVIKGAASPKSpezifikation
Antibody Production, Protein Array, ELISA, Western Blot | |
2558 | |
GABRA5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDNGMFSGFIMIKNLLLFCISMNLSSHFGFSQMPTSSVKDETNDNITIFTRILDGLLDGYDNRLRPGLGERITQVRTDIYVTSFGPVSDTEMEYTIDVFFRQSWKDERLRFKGPMQRLPLNNLLASKIWTPDTFFHNGKKSIAHNMTTPNKLPRLEDDGTLLYTMRLTISAECPMQLEDFPMDAHACPLKFGSYAYPNSEVVYVWTNGSTKSVVVAEDGSRLNQYHLMGQTVGTENISTSTGEYTIMTAHFHLKRKIGYFVIQTYLPCIMTVILSQVSFWLNRESVPARTVFGVTTVLTMTTLSISARNSLPKVAYATAMDWFIAVCYAFVFSALIEFATVNYFTKRGWAWDGKKALEAAKIKKKREVILNKSTNAFTTGKMSHPPNIPKEQTPAGTSNTTSVSVKPSEEKTSESKKTYNSISKIDKMSRIVFPVLFGTFNLVYWATYLNREPVIKGAASPK | |
RUO | |
GABRA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.56kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC138184 | |
GABRA5 | |
Recombinant | |
wheat germ expression system |