missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GDAP1L1 Full-length ORF (NP_076939.3, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00078997-P01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16163983

  • 335.00 € / 10 Mikrogramm
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. [provided by RefSeq]

Sequence: MATPNNLTPTNCSWWPISALESDAAKPAEAPDAPEAASPAHWPRESLVLYHWTQSFSSQKVRLVIAEKGLVCEERDVSLPQSEHKEPWFMRLNLGEEVPVIIHRDNIISDYDQIIDYVERTFTGEHVVALMPEVGSLQHARVLQYRELLDALPMDAYTHGCILHPELTTDSMIPKYATAEIRRHLANATTDLMKLDHEEEPQLSEPYLSKQKKLMAKILEHDDVSYLKKILGELAMVLDQIEAELEKRKLENEGQKCELWLCGCAFTLADVLLGATLHRLKFLGLSKKYWEDGSRPNLQSFFERVQRRFAFRKVLGDIHTTLLSAVIPNAFRLVKRKPPSFFGASFLMGSLGGMGYFAYWYLKKKYI
Spezifikation

Spezifikation

NP_076939.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp761K228/dJ881L22.1/dJ995J12.1.1
GDAP1L1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
78997
GDAP1L1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATPNNLTPTNCSWWPISALESDAAKPAEAPDAPEAASPAHWPRESLVLYHWTQSFSSQKVRLVIAEKGLVCEERDVSLPQSEHKEPWFMRLNLGEEVPVIIHRDNIISDYDQIIDYVERTFTGEHVVALMPEVGSLQHARVLQYRELLDALPMDAYTHGCILHPELTTDSMIPKYATAEIRRHLANATTDLMKLDHEEEPQLSEPYLSKQKKLMAKILEHDDVSYLKKILGELAMVLDQIEAELEKRKLENEGQKCELWLCGCAFTLADVLLGATLHRLKFLGLSKKYWEDGSRPNLQSFFERVQRRFAFRKVLGDIHTTLLSAVIPNAFRLVKRKPPSFFGASFLMGSLGGMGYFAYWYLKKKYI
RUO
GDAP1L1
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote