missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GIMAP5 Partial ORF (NP_060854.2, 186 a.a. - 284 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Zugriffsnummer NP_060854.2
Zur Verwendung mit (Anwendung) Antibody Production, ELISA, Protein Array, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 55340
Molekulargewicht 36.63kDa
View More Specs

Products 2
Product Code Brand Menge Price Quantity & Availability  
Product Code Brand Menge Price Quantity & Availability  
16168086
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00055340-Q01.10UG
10 ug
335.00 €
10 Mikrogramm
View alternative products
missing translation for 'toSaveUpto' 
»
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
16178086
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00055340-Q01.25UG
25 ug
508.00 €
25 Mikrogramm
View alternative products
missing translation for 'toSaveUpto' 
»
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description

Description

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1. [provided by RefSeq]

Sequence: AFNNWGSVEEQRQQQAELLAVIERLGREREGSFHSNDLFLDAQLLQRTGAGACQEDYRQYQAKVEWQVEKHKQELRENESNWAYKALLRVKHLMLLHYE
Specifications

Specifications

NP_060854.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11296/HIMAP3/IAN4/IAN4L1/IAN5/IMAP3/hIAN5
GIMAP5
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
55340
GIMAP5 (Human) Recombinant Protein (Q01)
AFNNWGSVEEQRQQQAELLAVIERLGREREGSFHSNDLFLDAQLLQRTGAGACQEDYRQYQAKVEWQVEKHKQELRENESNWAYKALLRVKHLMLLHYE
RUO
GIMAP5
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates