Learn More
ACROBiosystems Human Glypican 3 / GPC3 (S359F) Protein, His Tag, low endotoxin
low endotoxin
Marke: ACROBiosystems GP3-H5223-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.7 kDa. The protein migrates as 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH35972.1 | |
100 μg | |
Gln 25 - His 559 | |
GPC3,OCI5,Glypican-3,GTR2-2,MXR7,DGSX,SDYS ,SGB,SGBS,SGBS1 | |
Unconjugated | |
His Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
61.7 kDa | |
Human Glypican 3 (S359F), His Tag, low endotoxin (GP3-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1 (S359F)). | |
-20°C | |
Glypican 3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |