missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GNAT2 Full-length ORF (NP_005263.1, 1 a.a. - 354 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00002780-P01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16168297

  • 482.88 € / 25 Mikrogramm

Voraussichtliches Versanddatum: 10-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in cones. [provided by RefSeq]

Sequence: MGSGASAEDKELAKRSKELEKKLQEDADKEAKTVKLLLLGAGESGKSTIVKQMKIIHQDGYSPEECLEFKAIIYGNVLQSILAIIRAMTTLGIDYAEPSCADDGRQLNNLADSIEEGTMPPELVEVIRRLWKDGGVQACFERAAEYQLNDSASYYLNQLERITDPEYLPSEQDVLRSRVKTTGIIETKFSVKDLNFRMFDVGGQRSERKKWIHCFEGVTCIIFCAALSAYDMVLVEDDEVNRMHESLHLFNSICNHKFFAATSIVLFLNKKDLFEEKIKKVHLSICFPEYDGNNSYDDAGNYIKSQFLDLNMRKDVKEIYSHMTCATDTQNVKFVFDAVTDIIIKENLKDCGLF
Spezifikation

Spezifikation

NP_005263.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACHM4/GNATC
GNAT2
Yes
wheat germ expression system
Liquid
2780
GNAT2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSGASAEDKELAKRSKELEKKLQEDADKEAKTVKLLLLGAGESGKSTIVKQMKIIHQDGYSPEECLEFKAIIYGNVLQSILAIIRAMTTLGIDYAEPSCADDGRQLNNLADSIEEGTMPPELVEVIRRLWKDGGVQACFERAAEYQLNDSASYYLNQLERITDPEYLPSEQDVLRSRVKTTGIIETKFSVKDLNFRMFDVGGQRSERKKWIHCFEGVTCIIFCAALSAYDMVLVEDDEVNRMHESLHLFNSICNHKFFAATSIVLFLNKKDLFEEKIKKVHLSICFPEYDGNNSYDDAGNYIKSQFLDLNMRKDVKEIYSHMTCATDTQNVKFVFDAVTDIIIKENLKDCGLF
RUO
GNAT2
Wheat Germ (in vitro)
GST
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate