Learn More
Abnova™ Human GPR62 Partial ORF (NP_543141, 314 a.a. - 368 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00118442-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.[supplied by OMIM]
Sequence: RACTPQAWHPRALLQCLQRPPEGPAVGPSEAPEQTPELAGGRSPAYQGPPESSLSSpezifikation
NP_543141 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.79kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RACTPQAWHPRALLQCLQRPPEGPAVGPSEAPEQTPELAGGRSPAYQGPPESSLS | |
RUO | |
GPR62 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
118442 | |
GPR62 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPCR8/KPG_005/MGC26943 | |
GPR62 | |
Recombinant | |
wheat germ expression system |