Learn More
Abnova™ Human GRHL2 Full-length ORF (NP_079191.1, 1 a.a. - 625 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00079977-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of non-syndromic sensorineural deafness autosomal dominant type 28 (DFNA28)
Sequence: MSQESDNNKRLVALVPMPSDPPFNTRRAYTSEDEAWKSYLENPLTAATKAMMSINGDEDSAAALGLLYDYYKVPRDKRLLSVSKASDSQEDQEKRNCLGTSEAQSNLSGGENRVQVLKTVPVNLSLNQDHLENSKREQYSISFPESSAIIPVSGITVVKAEDFTPVFMAPPVHYPRGDGEEQRVVIFEQTQYDVPSLATHSAYLKDDQRSTPDSTYSESFKDAATEKFRSASVGAEEYMYDQTSSGTFQYTLEATKSLRQKQGEGPMTYLNKGQFYAITLSETGDNKCFRHPISKVRSVVMVVFSEDKNRDEQLKYWKYWHSRQHTAKQRVLDIADYKESFNTIGNIEEIAYNAVSFTWDVNEEAKIFITVNCLSTDFSSQKGVKGLPLMIQIDTYSYNNRSNKPIHRAYCQIKVFCDKGAERKIRDEERKQNRKKGKGQASQTQCNSSSDGKLAAIPLQKKSDITYFKTMPDLHSQPVLFIPDVHFANLQRTGQVYYNTDDEREGGSVLVKRMFRPMEEEFGPVPSKQMKEEGTKRVLLYVRKETDDVFDALMLKSPTVKGLMEAISEKYGLPVEKIAKLYKKSKKGILVNMDDNIIEHYSNEDTFILNMESMVEGFKVTLMEISpezifikation
NP_079191.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
97.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BOM/DFNA28/FLJ11172/FLJ13782/MGC149294/MGC149295/TFCP2L3 | |
GRHL2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79977 | |
GRHL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSQESDNNKRLVALVPMPSDPPFNTRRAYTSEDEAWKSYLENPLTAATKAMMSINGDEDSAAALGLLYDYYKVPRDKRLLSVSKASDSQEDQEKRNCLGTSEAQSNLSGGENRVQVLKTVPVNLSLNQDHLENSKREQYSISFPESSAIIPVSGITVVKAEDFTPVFMAPPVHYPRGDGEEQRVVIFEQTQYDVPSLATHSAYLKDDQRSTPDSTYSESFKDAATEKFRSASVGAEEYMYDQTSSGTFQYTLEATKSLRQKQGEGPMTYLNKGQFYAITLSETGDNKCFRHPISKVRSVVMVVFSEDKNRDEQLKYWKYWHSRQHTAKQRVLDIADYKESFNTIGNIEEIAYNAVSFTWDVNEEAKIFITVNCLSTDFSSQKGVKGLPLMIQIDTYSYNNRSNKPIHRAYCQIKVFCDKGAERKIRDEERKQNRKKGKGQASQTQCNSSSDGKLAAIPLQKKSDITYFKTMPDLHSQPVLFIPDVHFANLQRTGQVYYNTDDEREGGSVLVKRMFRPMEEEFGPVPSKQMKEEGTKRVLLYVRKETDDVFDALMLKSPTVKGLMEAISEKYGLPVEKIAKLYKKSKKGILVNMDDNIIEHYSNEDTFILNMESMVEGFKVTLMEI | |
RUO | |
GRHL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |