missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IFN-alpha 2 (alpha2a) Recombinant Protein, PeproTech®

Artikelnummer. 17836998
Change view
Click to view available options
Menge:
1 mg
100 μg
20 μg
250 μg
500 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
20 Mikrogramm
250 Mikrogramm
500 Mikrogramm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17836998 20 μg 20 Mikrogramm
17826768 100 μg 100 Mikrogramm
17896898 1 mg 1 Milligramm
17886998 250 μg 250 Mikrogramm
17856828 500 μg 500 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17836998 Lieferant Gibco™ Lieferanten-Nr. 30002AA20UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Recombinant Human IFN-alpha (2a) is a 19.3 kDa protein containing 166 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P01563
Zur Verwendung mit (Anwendung) Functional Assay
Format Lyophilized
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 3440
Molekulargewicht 19.3 kDa
Name Human IFN-alpha 2 (alpha2a)
Reinigungsverfahren HPLC, SDS-PAGE
Menge 20 μg
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/ μg
Gen-Alias alpha-2 A interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1 b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1 b; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2
Gebräuchliche Bezeichnung Interferon alpha-2
Gensymbol IFNA2
Biologische Aktivität Determined by a cytotoxicity assay using human TF-1 cells.
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz MCDLPQTHSL GSRRTLMLLA QMRKISLFSC LKDRHDFGFP QEEFGNQFQK AETIPVLHEM IQQIFNLFST KDSSAAWDET LLDKFYTELY QQLNDLEACV IQGVGVTETP LMKEDSILAV RKYFQRITLY LKEKKYSPCA WEVVRAEIMR SFSLSTNLQE SLRSKE
Inhalt und Lagerung -20°C
Aktivität Determined by a cytotoxicity assay using human TF-1 cells.
Expressionssystem E. coli
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.