Learn More
R&D Systems™ Human IFN-alpha Multi-Subtype ELISA Kit, R&D Systems™
Human IFN-alpha Multi-Subtype ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marke:  R&D Systems™ 41105-1
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13414174
  • 977.87 € / 96 Tests
Voraussichtlicher Liefertermin 30-07-2019
Zum Warenkorb hinzufügen
Beinhaltet:
  • Pre-coated microtiter plate(s)
  • Plate sealers
  • Wash Solution Concentrate
  • Human Interferon Alpha Standard, 10,000 pg/ml
  • Dilution Buffer
  • Antibody Concentrate
  • HRP Conjugate Concentrate
  • Con
Beschreibung
Beschreibung
The Human IFN-alpha Multi-Subtype ELISA Kit quantifies human IFN-alpha in cell culture supernates (100 ul).
Spezifikation
Spezifikation
Biotin
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates
Human
Store the unopened product at 2 - 8° C. Do not use past expiration date.
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3440
1 Plate
12.5pg/mL
Cell Culture Supernates (100μL)
Human IFN-α
Sufficient materials to run ELISAs on at least one 96-well plate
Videos