Learn More
R&D Systems™ Human IFN-alpha Multi-Subtype ELISA Kit, R&D Systems™
formulated to detect 14 out of 15 identified human IFN-α subtypes
Marke:  R&D Systems™ 41105-1
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13414174
  • 977.87 € / 96 Tests
Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen
Beinhaltet:
  • Pre-coated microtiter plate(s)
  • Plate sealers
  • Wash Solution Concentrate
  • Human Interferon Alpha Standard, 10,000 pg/ml
  • Dilution Buffer
  • Antibody Concentrate
  • HRP Conjugate Concentrate
  • Con
Beschreibung
Beschreibung
VeriKine™ Human IFN-α Multi-Subtype ELISA kit is specifically formulated to detect 14 out of 15 identified human IFN-α subtypes. They are: IFN-αA, IFN-α2, IFN-αD, IFN-αB2, IFN-αC, IFN-αG, IFN-αH, IFN-αI, IFN-αJ1, IFN-αK, IFN-α1, IFN-α4a, IFN-α4b, and IFN-αWA.
Spezifikation
Spezifikation
Biotin
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates
Human
Store the unopened product at 2 - 8° C. Do not use past expiration date.
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3440
1 Plate
12.5pg/mL
Cell Culture Supernates (100μL)
Human IFN-α
Sufficient materials to run ELISAs on at least one 96-well plate
Videos