missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IFN-beta, Animal-Free Recombinant Protein, PeproTech®

Artikelnummer. 17827078
Change view
Click to view available options
Menge:
250 μg
5 μg
Packungsgröße:
250 Mikrogramm
5 Mikrogramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17827078 250 μg 250 Mikrogramm
17826508 5 μg 5 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17827078 Lieferant Gibco™ Lieferanten-Nr. AF30002B250UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

PeproTech's Recombinant Human IFN-beta is a 20.0 kDa protein containing 166 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P01574
Zur Verwendung mit (Anwendung) Functional Assay
Format Lyophilized
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 3456
Molekulargewicht 20 kDa
Name Human IFN-beta, Animal-Free
Reinigungsverfahren HPLC, SDS-PAGE
Menge 250 μg
Kennzeichnung RUO
Endotoxin-Konzentration <0.1 EU/ μg
Gen-Alias Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1
Gebräuchliche Bezeichnung IFN beta
Gensymbol Ifnb1
Biologische Aktivität Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤ 0.25 ng/ml, corresponding to a specific activity of ≥ 4 x 10^6 units/mg
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz MSYNLLGFLQ RSSNFQCQKL LWQLNGRLEY CLKDRMNFDI PEEIKQLQQF QKEDAALTIY EMLQNIFAIF RQDSSSTGWN ETIVENLLAN VYHQINHLKT VLEEKLEKED FTRGKLMSSL HLKRYYGRIL HYLKAKEYSH CAWTIVRVEI LRNFYFINRL TGYLRN
Inhalt und Lagerung -20°C
Aktivität Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤ 0.25 ng/ml, corresponding to a specific activity of ≥ 4 x 10^6 units/mg
Expressionssystem E. coli
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.