Learn More
Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Artikelnummer. p-7250044
Change view
Click to view available options
Menge:
100 μg
25 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18750067 25 μg 25 Mikrogramm
18791447 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18750067 Lieferant R&D Systems Lieferanten-Nr. MAB11127SP
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
IFN-beta Monoclonal antibody specifically detects IFN-beta in Human samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen IFN-beta
Anwendungen Neutralization
Klassifikation Monoclonal
Klon 1020369
Konjugat Unconjugated
Verdünnung Neutralization
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gen-Zugriffsnummer P01574
Gen-Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Wirtsspezies Mouse
Immunogen CHO-expressed recombinant Human IFN-beta protein, Met22-Asn187, Accession # P01574
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 25 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 3456
Rekonstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1