missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgA Isotype Control

Artikelnummer. 11865895
Change view
Click to view available options
Menge:
2 mg
Packungsgröße:
2 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
11865895 2 mg 2 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11865895 Lieferant Invitrogen™ Lieferanten-Nr. 31148

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Human Polyclonal Isotype Control

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity: The precipitin lines against goat anti-human whole serum are the same as those against goat anti-human IgA, alpha chain specific, when assayed by immunoelectrophoresis at an antigen concentration of 20 mg/mL. No reaction was observed against goat anti-human IgG, Fc fragment-specific or goat anti-human IgM, Fc5-mu fragment specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Storage: Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of Origin: USA.

Human IgA, also known as human immunoglobulin A, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in mucosal secretions, such as saliva, tears, and breast milk, and plays a key role in defense against pathogens at mucosal surfaces. IgA works by binding to specific antigens and preventing them from entering the body's tissues. It can also activate complement and promote phagocytosis by immune cells. Human IgA can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious diseases.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Human IgA
Anwendungen Control, Inhibition Assays, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry
Klassifikation Polyclonal
Konzentration 4.49 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative; pH 7.6
Gen-Alias A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker)
Wirtsspezies Human
Reinigungsverfahren Purified
Menge 2 mg
Regulatorischer Status RUO
Primär oder sekundär Isotype Control (Primary)
Zielspezies Not Applicable
Inhalt und Lagerung 4°C
Produkttyp Isotype Control
Form Liquid
Isotype IgA
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.