missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IL-1 RAcP / IL-1 R3 Protein, Fc Tag

Marke:  ACROBiosystems ILP-H5256-1MG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15897107

  • 1307.63 € / 1 Milligramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Interleukin-1 receptor accessory protein (IL1RAP or IL-1RAcP) is also known as Interleukin-1 receptor 3 (IL-1R-3 or IL-1R3), C3orf13, which belongs to the interleukin-1 receptor family. IL1RAP contains three Ig-like C2-type (immunoglobulin-like) domains and one TIR domain. IL1RAP is detected in liver, skin, placenta, thymus and lung. IL-1R3 / IL1RAP is coreceptor with IL1R1, associates with IL1R1 bound to IL1B to form the high affinity interleukin-1 receptor complex which mediates interleukin-1-dependent activation of NF-kappa-B and other pathways.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 65.7 kDa. The protein migrates as 75-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

65.7 kDa
Human IL-1 RAcP, Fc Tag (ILP-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Glu 359 (Accession # Q9NPH3).
-20°C
IL-1 RAcP
Human
50 mM Tris, 100 mM Glycine, pH7.5
HEK293
Bio-Markers and CD Antigens
1 mg
Ser 21 - Glu 359
IL1RAP,IL-1 RAcP,C3orf13,IL1R3
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote