missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-10 Quantikine ELISA Kit, R&D Systems™

a 3.5 or 4.5 hour solid phase ELISA designed to measure IL-10 in cell culture supernates, serum, and plasma.

Marke:  R&D Systems™ S1000B

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412646

  • 4113.82 € / 6 Stück


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains Sf 21-expressed recombinant human IL-10 and antibodies raised against the recombinant factor. This immunoassay has been shown to accurately quantitate recombinant human IL-10 and does not cross-react with viral IL-10. Results obtained using natural human IL-10 showed linear curves that were parallel to the standard curves obtained using the recombinant Quantikine kit standards. These results indicate this kit can be used to determine relative mass values for natural human IL-10
Spezifikation

Spezifikation

7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
3586
6 Plates
3.9pg/mL
Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Natural and recombinant human IL-10
Sufficient materials to run ELISAs on at least six 96-well plates
HRP
ELISA
Solid Phase Sandwich ELISA
1/Pk.
Cell Culture Supernates (200uL), Serum (200uL), EDTA Plasma (200uL), Heparin Plasma (200uL), Citrate Plasma (200uL)
Human
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos