missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-10 Quantikine HS ELISA Kit, R&D Systems™

a 5.5 hour solid phase ELISA designed to measure IL-10 in serum and plasma.

Marke:  R&D Systems™ SS100C

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412876

  • 5087.38 € / 6 Stück


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains Sf 21-expressed recombinant human IL-10 and antibodies raised against the recombinant factor. This immunoassay has been shown to accurately quantitate recombinant human IL-10. Results obtained using natural human IL-10 showed linear curves that are parallel to the standard curves obtained using the recombinant kit standards. These results indicate that this kit can be used to determine relative mass values for natural human IL-10
Spezifikation

Spezifikation

0.78 - 50pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
3586
6 Plates
0.17pg/mL
Serum (200μL), EDTA Plasma (200μL), Citrate Plasma (200μL)
Natural and recombinant human IL-10
Sufficient materials to run ELISAs on at least six 96-well plates
HRP
ELISA
Solid Phase Sandwich ELISA
1/Pk.
Serum (200uL), EDTA Plasma (200uL), Citrate Plasma (200uL)
Human
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos