Learn More
R&D Systems™ Human IL-10 Quantikine HS ELISA Kit, R&D Systems™
Human IL-10 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich | 0.78 - 50pg/mL |
---|---|
Assay-Empfindlichkeit: | 0.17pg/mL |
Probentyp | Plasma (EDTA, Citrate), Serum |
Beschreibung
The Human IL-10 Quantikine HS ELISA Kit quantifies human IL-10 in serum (200 ul), edta plasma (200 ul), citrate plasma (200 ul).Spezifikation
0.78 - 50pg/mL | |
0.17pg/mL | |
Plasma (EDTA, Citrate), Serum |