missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha L/CD11a Antibody, R&D Systems™

Beschreibung
Integrin alpha L/CD11a Monoclonal specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
Specifications
| Antigen | Integrin alpha L/CD11a |
| Anwendungen | Flow Cytometry, CyTOF |
| Klassifikation | Monoclonal |
| Klon | CR38 |
| Konzentration | LYOPH |
| Konjugat | Unconjugated |
| Verdünnung | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gen-Alias | CD11 antigen-like family member A, CD11a, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), ITGAL, Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, Ly-15, Ly-21, lymphocyte function-associated antigen 1 |
| Gensymbole | ITGAL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?