Learn More
ACROBiosystems Human Integrin alpha V beta 3 (ITGAV&ITGB3) Heterodimer Protein, His Tag&Tag Free
Marke:  ACROBiosystems IT3-H52E3-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15887147
  • 3838.91 € / 1 Milligramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Integrin alpha-V/beta-3 (ITGAV:ITGB3) is a receptor for cytotactin, fibronectin, laminin, matrix metalloproteinase-2, osteopontin, osteomodulin, prothrombin, thrombospondin, vitronectin and von Willebrand factor. Integrins alpha-IIb/beta-3 and alpha-V/beta-3 recognize the sequence R-G-D in a wide array of ligands. Also, Integrin alpha-V/beta-3 acts as a receptor for herpes virus 8/HHV-8, coxsackievirus A9, Hantaan virus, cytomegalovirus/HHV-5, human metapneumovirus, human parechovirus 1 and west nile virus. Furthermore, in case of HIV-1 infection, the interaction with extracellular viral Tat protein seems to enhance angiogenesis in Kaposi's sarcoma lesions.
Human ITGAV&ITGB3 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAV and ITGB3, has a calculated MW of 112.9 kDa (ITGAV) and 81.8 kDa (ITGB3). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistid
Spezifikation
Spezifikation
NP_002201.1 (ITGAV) and NP_000203.2 (ITGB3)
1 mg
Phe 31 - Val 992 (ITGAV) and Gly 27 - Asp 718 (ITGB3)
Integrin alpha V beta 3,ITGAVandITGB3
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
112.9 kDa (ITGAV) and 81.8 kDa (ITGB3)
Human ITGAV&ITGB3; Heterodimer Protein, His Tag&Tag; Free (IT3-H52E3) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) and Gly 27 - Asp 718 (ITGB3) (Accession # NP_002201.1 (ITGAV) and NP_000203.2 (ITGB3)).
-20°C
Integrin alpha V beta 3
Human
50 mM Tris, 150 mM NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens