Learn More
Abnova™ Human ITGA6 Partial ORF (NP_000201, 24 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00003655-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The ITGA6 protein product is the integrin alpha chain alpha 6. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: FNLDTREDNVIRKYGDPGSLFGFSLAMHWQLQPEDKRLLLVGAPRAEALPLQRANRTGGLYSCDITARGPCTRIEFDNDADPTSESKEDQWMGVTVQSQGPGGKVVTCAHSpezifikation
NP_000201 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FNLDTREDNVIRKYGDPGSLFGFSLAMHWQLQPEDKRLLLVGAPRAEALPLQRANRTGGLYSCDITARGPCTRIEFDNDADPTSESKEDQWMGVTVQSQGPGGKVVTCAH | |
RUO | |
ITGA6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3655 | |
ITGA6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD49f/DKFZp686J01244/FLJ18737/ITGA6B/VLA-6 | |
ITGA6 | |
Recombinant | |
wheat germ expression system |