Learn More
Abnova™ Human ITGB1BP1 Full-length ORF (AAH12264, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009270-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The cytoplasmic domains of integrins are essential for cell adhesion. The protein encoded by this gene binds to the beta1 integrin cytoplasmic domain. The interaction between this protein and beta1 integrin is highly specific. Two isoforms of this protein are derived from alternatively spliced transcripts. The shorter form of this protein does not interact with the beta1 integrin cytoplasmic domain. The longer form is a phosphoprotein and the extent of its phosphorylation is regulated by the cell-matrix interaction, suggesting an important role of this protein during integrin-dependent cell adhesion. [provided by RefSeq]
Sequence: MFRKGKKRHSSSSSQSSEISTKSKSVDSSLGGLSRSSTVASLDTDSTKSSGQSNNNSDTCAEFRIKYVGAIEKLKLSEGKGLEGPLDLINYIDVAQQDGKLPFVPPEEEFIMGVSKYGIKVSTSDQYDVLHRHALYLIIRMVCYDDGLGVGKSLLALKTTDASNEEYSLWVYQCNSLEQAQAICKVLSTAFDSVLTSEKPSpezifikation
AAH12264 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.74kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686K08158/ICAP-1A/ICAP-1B/ICAP-1alpha/ICAP1/ICAP1A/ICAP1B | |
ITGB1BP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9270 | |
ITGB1BP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFRKGKKRHSSSSSQSSEISTKSKSVDSSLGGLSRSSTVASLDTDSTKSSGQSNNNSDTCAEFRIKYVGAIEKLKLSEGKGLEGPLDLINYIDVAQQDGKLPFVPPEEEFIMGVSKYGIKVSTSDQYDVLHRHALYLIIRMVCYDDGLGVGKSLLALKTTDASNEEYSLWVYQCNSLEQAQAICKVLSTAFDSVLTSEKP | |
RUO | |
ITGB1BP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |