Learn More
Abnova™ Human KRT25 Full-length ORF (AAI46489.1, 1 a.a. - 450 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00147183-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the type I (acidic) keratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. The type I keratin genes are clustered in a region of chromosome 17q12-q21. [provided by RefSeq]
Sequence: MSLRLSSASRRSCPRPTTGSLRLYGGGTSFGTGNSCGISGIGSGFSSAFGGSSSGGNTGGGNPCAGFTVNERGLLSGNEKVTMQNLNDRLASYLDSVHALEEANADLEQKIKGWYEKFGPGSCRGLDHDYSRYFPIIDDLKNQIIASTTSNANAVLQIDNARLTADDFRLKYENELALHQSVEADVNGLRRVLDEITLCRTDLEIQYETLSEEMTYLKKNHKEEMQVLQCAAGGNVNVEMNAAPGVDLTVLLNNMRAEYEALAEQNRRDAEAWFNEKSASLQQQISEDVGATTSARNELTEMKRTLQTLEIELQSLLATKHSLECSLTETESNYCAQLAQIQAQIGALEEQLHQVRTETEGQKLEYEQLLDIKLHLEKEIETYCLLIGGDDGACKSGGYKSKDYGSGNVGSQVKDPAKAIVVKKVLEEVDQRSKILTTRLHSLEEKSQSNSpezifikation
AAI46489.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.45kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRT25A | |
KRT25 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
147183 | |
KRT25 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSLRLSSASRRSCPRPTTGSLRLYGGGTSFGTGNSCGISGIGSGFSSAFGGSSSGGNTGGGNPCAGFTVNERGLLSGNEKVTMQNLNDRLASYLDSVHALEEANADLEQKIKGWYEKFGPGSCRGLDHDYSRYFPIIDDLKNQIIASTTSNANAVLQIDNARLTADDFRLKYENELALHQSVEADVNGLRRVLDEITLCRTDLEIQYETLSEEMTYLKKNHKEEMQVLQCAAGGNVNVEMNAAPGVDLTVLLNNMRAEYEALAEQNRRDAEAWFNEKSASLQQQISEDVGATTSARNELTEMKRTLQTLEIELQSLLATKHSLECSLTETESNYCAQLAQIQAQIGALEEQLHQVRTETEGQKLEYEQLLDIKLHLEKEIETYCLLIGGDDGACKSGGYKSKDYGSGNVGSQVKDPAKAIVVKKVLEEVDQRSKILTTRLHSLEEKSQSN | |
RUO | |
KRT25 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |