missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LAT2 Partial ORF (NP_071323.1, 144 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00007462-Q01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16167515

  • 320.54 € / 10 Mikrogramm

Voraussichtliches Versanddatum: 14-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene is one of the contiguous genes at 7q11.23 commonly deleted in Williams syndrome, a multisystem developmental disorder. This gene consists of at least 14 exons, and its alternative splicing generates 3 transcript variants, all encoding the same protein. [provided by RefSeq]

Sequence: QKTTETGAQQEGIGGLCRGDLSLSLALKTGPTSGLCPSASPEEDEESEDYQNSASIHQWRESRKVMGQLQREASPGPVGSPDEEDGEPDYVNGEVAATE
Spezifikation

Spezifikation

NP_071323.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QKTTETGAQQEGIGGLCRGDLSLSLALKTGPTSGLCPSASPEEDEESEDYQNSASIHQWRESRKVMGQLQREASPGPVGSPDEEDGEPDYVNGEVAATE
RUO
LAT2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
7462
LAT2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSPC046/LAB/NTAL/WBSCR15/WBSCR5/WSCR5
LAT2
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate