Learn More
Abnova™ Human LTBP2 Partial ORF (NP_000419, 1709 a.a. - 1818 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004053-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene belongs to the family of latent transforming growth factor (TGF)-beta binding proteins (LTBP), which are extracellular matrix proteins with multi-domain structure. This protein is the largest member of the LTBP family possessing unique regions and with most similarity to the fibrillins. It has thus been suggested that it may have multiple functions: as a member of the TGF-beta latent complex, as a structural component of microfibrils, and a role in cell adhesion. [provided by RefSeq]
Sequence: LQPSELQPHYVASHPEPPAGFEGLQAEECGILNGCENGRCVRVREGYTCDCFEGFQLDAAHMACVDVNECDDLNGPAVLCVHGYCENTEGSYRCHCSPGYVAEAGPPHCTSpezifikation
NP_000419 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LQPSELQPHYVASHPEPPAGFEGLQAEECGILNGCENGRCVRVREGYTCDCFEGFQLDAAHMACVDVNECDDLNGPAVLCVHGYCENTEGSYRCHCSPGYVAEAGPPHCT | |
RUO | |
LTBP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4053 | |
LTBP2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C14orf141/LTBP3/MSTP031 | |
LTBP2 | |
Recombinant | |
wheat germ expression system |