missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MFSD2 Full-length ORF (NP_116182.2, 1 a.a. - 530 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084879-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MAKGEGAESGSAAGLLPTSILQSTERPAQVKKEPKKKKQQLSVCNKLCYALGGAPYQVTGCALGFFLQIYLLDVAQVGPFSASIILFVGRAWDAITDPLVGLCISKSPWTCLGRLMPWIIFSTPLAVIAYFLIWFVPDFPHGQTYWYLLFYCLFETMVTCFHVPYSALTMFISTEQTERDSATAYRMTVEVLGTVLGTAIQGQIVGQADTPCFQDLNSSTVASQSANHTHGTTSHRETQKAYLLAAGVIVCIYIICAVILILGVREQREPYEAQQSEPIAYFRGLRLVMSHGPYIKLITGFLFTSLAFMLVEGNFVLFCTYTLGFRNEFQNLLLAIMLSATLTIPIWQWFLTRFGKKTAVYVGISSAVPFLILVALMESNLIITYAVAVAAGISVAAAFLLPWSMLPDVIDDFHLKQPHFHGTEPIFFSFYVFFTKFASGVSLGISTLSLDFAGYQTRGCSQPERVKFTLNMLVTMAPIVLILLGLLLFKMYPIDEERRRQNKKALQALRDEASSSGCSETDSTELASILSpezifikation
NP_116182.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
85kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14490/FLJ35904/FP1147/HMFN0656/PP10484/RP3-342P20.1 | |
MFSD2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84879 | |
MFSD2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKGEGAESGSAAGLLPTSILQSTERPAQVKKEPKKKKQQLSVCNKLCYALGGAPYQVTGCALGFFLQIYLLDVAQVGPFSASIILFVGRAWDAITDPLVGLCISKSPWTCLGRLMPWIIFSTPLAVIAYFLIWFVPDFPHGQTYWYLLFYCLFETMVTCFHVPYSALTMFISTEQTERDSATAYRMTVEVLGTVLGTAIQGQIVGQADTPCFQDLNSSTVASQSANHTHGTTSHRETQKAYLLAAGVIVCIYIICAVILILGVREQREPYEAQQSEPIAYFRGLRLVMSHGPYIKLITGFLFTSLAFMLVEGNFVLFCTYTLGFRNEFQNLLLAIMLSATLTIPIWQWFLTRFGKKTAVYVGISSAVPFLILVALMESNLIITYAVAVAAGISVAAAFLLPWSMLPDVIDDFHLKQPHFHGTEPIFFSFYVFFTKFASGVSLGISTLSLDFAGYQTRGCSQPERVKFTLNMLVTMAPIVLILLGLLLFKMYPIDEERRRQNKKALQALRDEASSSGCSETDSTELASIL | |
RUO | |
MFSD2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |