missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MGC13096 Partial ORF (AAH06146, 80 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084306-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: CACPGCSTGGARSWKVFRSQCLQVPEREAQDAQKQGNSLAAEDWCEGADDWGSDTEEGPSPQFTLDFGNDASSAKDVDWTARLQDLRLQDAVLGAAHPVPSpezifikation
AAH06146 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CACPGCSTGGARSWKVFRSQCLQVPEREAQDAQKQGNSLAAEDWCEGADDWGSDTEEGPSPQFTLDFGNDASSAKDVDWTARLQDLRLQDAVLGAAHPVP | |
RUO | |
PDCD2L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84306 | |
MGC13096 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC13096 | |
PDCD2L | |
Recombinant | |
wheat germ expression system |