missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human MMP-9 Quantikine ELISA Kit, R&D Systems™

a 3.5 hour solid phase ELISA designed to measure MMP-9 (92 kDa pro- and 82 kDa active forms but not the 65 kDa form) in cell culture supernates, saliva, serum, plasma, and urine.

Marke:  R&D Systems™ SMP900

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412836

  • 4113.82 € / 6 Stück


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It is calibrated with CHO cell-expressed recombinant human pro-MMP-9 and the antibodies were raised against the recombinant factor. Natural human MMP-9 showed dose-response curves that were parallel to the standard curves obtained using the recombinant Quantikine kit standards, indicating that this kit can be used to determine relative mass values of natural human MMP-9
Spezifikation

Spezifikation

0.31 - 20ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
4318
1 Pk.
Cell Culture Supernates (10uL), Serum (10uL), Platelet-poor Heparin Plasma (10uL), Saliva (10uL), Urine (100uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
HRP
ELISA
6 Plates
0.156ng/mL
Cell Culture Supernates (10μL), Serum (10μL), Platelet-poor Heparin Plasma (10μL), Saliva (10μL), Urine (100μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos