Learn More
Abnova™ Human MMP21 Partial ORF (NP_671724.1, 374 a.a. - 481 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00118856-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the matrix metalloproteinase family. Proteins in this family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction, and tissue remodeling,and disease processes, such as asthma and metastasis. The encoded protein may play an important role in embryogenesis, particularly in neuronal cells, as well as in lymphocyte development and survival. [provided by RefSeq]
Sequence: TRYGDPIQILTGWPGIPTHNIDAFVHIWTWKRDERYFFQGNQYWRYDSDKDQALTEDEQGKSYPKLISEGFPGIPSPLDTAFYDRRQKLIYFFKESLVFAFDVNRNRVSpezifikation
NP_671724.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TRYGDPIQILTGWPGIPTHNIDAFVHIWTWKRDERYFFQGNQYWRYDSDKDQALTEDEQGKSYPKLISEGFPGIPSPLDTAFYDRRQKLIYFFKESLVFAFDVNRNRV | |
RUO | |
MMP21 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
118856 | |
MMP21 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MMP21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |