missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MORC1 Partial ORF (NP_055244, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00027136-Q01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16185736

  • 335.00 € / 10 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes the human homolog of mouse morc and like the mouse protein it is testis-specific. Mouse studies support a testis-specific function since only male knockout mice are infertile; infertility is the only apparent defect. These studies further support a role for this protein early in spermatogenesis, possibly by affecting entry into apoptosis because testis from knockout mice show greatly increased numbers of apoptotic cells. [provided by RefSeq]

Sequence: MDDRYPALQRAQLRLDFIHANSTTHSFLFGALAELLDNARDAGAERLDVFSVDNEKLQGGFMLCFLDDGCGMSPEEASDIIYFGRSKKRLSTLKFIGQYG
Spezifikation

Spezifikation

NP_055244
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDDRYPALQRAQLRLDFIHANSTTHSFLFGALAELLDNARDAGAERLDVFSVDNEKLQGGFMLCFLDDGCGMSPEEASDIIYFGRSKKRLSTLKFIGQYG
RUO
MORC1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
27136
MORC1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MORC/ZCW6
MORC1
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote