missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse/Rat Phospho-JNK2 (T183/Y185) DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Marke:  R&D Systems™ DYC2236-2

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414455

  • 497.92 € / 2 Stück


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Spezifikation

Spezifikation

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
243911
1 Kit
Biotin
Two 96-well plates
ELISA Development Kit for phosphorylated intracellular proteins
Human, Mouse, Rat
Videos