missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human/Mouse/Rat Total Akt1 DuoSet IC ELISA
Human/Mouse/Rat Total Akt1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich | 312.00 to 20000 pg/mL |
---|---|
Kreuzreaktivität | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gen-ID (Entrez) | 207 |
Produkttyp | ELISA Development Kit for intracellular proteins |
Konjugat | Biotin |
Beschreibung
The Human/Mouse/Rat Total Akt1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat AKT1.Spezifikation
312.00 to 20000 pg/mL | |
207 | |
Biotin |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
Human, Mouse, Rat |