Learn More
R&D Systems™ Human/Mouse/Rat Total ERK2 DuoSet IC ELISA
Human/Mouse/Rat Total ERK2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Kreuzreaktivität | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
---|---|
Produkttyp | ELISA Development Kit for intracellular proteins |
Menge | 1 Kit (for 5 Plates) |
Spezies | Human, Mouse, Rat |
Beschreibung
The Human/Mouse/Rat Total ERK2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat ERK2.Spezifikation
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
1 Kit (for 5 Plates) | |
Human, Mouse, Rat |