Learn More
R&D Systems™ Human/Mouse/Rat Total ERK2 DuoSet IC ELISA
Human/Mouse/Rat Total ERK2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Kreuzreaktivität <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Produkttyp ELISA Development Kit for intracellular proteins
Menge 1 Kit (for 5 Plates)
Spezies Human, Mouse, Rat
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
The Human/Mouse/Rat Total ERK2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat ERK2.
Spezifikation
Spezifikation
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit (for 5 Plates)
Human, Mouse, Rat