missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human/Mouse Total Bcl-xL DuoSet IC, R&D Systems™
Human/Mouse Total Bcl-xL DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Kreuzreaktivität | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
---|---|
Produkttyp | ELISA Development Kit for intracellular proteins |
Menge | 1 Kit |
Gen-ID (Entrez) | 12048 |
Spezies | Human, Mouse |
Beschreibung
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously. Complete ELISpot kits are ready-to-r
The Human/Mouse Total Bcl-xL DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse Bcl-xL.Spezifikation
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
1 Kit | |
Human, Mouse |
ELISA Development Kit for intracellular proteins | |
12048 |