Learn More
Abnova™ Human MTMR2 Full-length ORF (NP_958435.1, 1 a.a. - 571 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008898-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is a member of the myotubularin family. The encoded protein possesses phosphatase activity towards phosphatidylinositol-3-phosphate and phosphatidylinositol-3,5-bisphosphate. Mutations in this gene are a cause of Charcot-Marie-Tooth disease type 4B, an autosomal recessive demyelinating neuropathy. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]
Sequence: MEEPPLLPGENIKDMAKDVTYICPFTGAVRGTLTVTNYRLYFKSMERDPPFVLDASLGVINRVEKIGGASSRGENSYGLETVCKDIRNLRFAHKPEGRTRRSIFENLMKYAFPVSNNLPLFAFEYKEVFPENGWKLYDPLLEYRRQGIPNESWRITKINERYELCDTYPALLVVPANIPDEELKRVASFRSRGRIPVLSWIHPESQATITRCSQPMVGVSGKRSKEDEKYLQAIMDSNAQSHKIFIFDARPSVNAVANKAKGGGYESEDAYQNAELVFLDIHNIHVMRESLRKLKEIVYPNIEETHWLSNLESTHWLEHIKLILAGALRIADKVESGKTSVVVHCSDGWDRTAQLTSLAMLMLDGYYRTIRGFEVLVEKEWLSFGHRFQLRVGHGDKNHADADRSPVFLQFIDCVWQMTRQFPTAFEFNEYFLITILDHLYSCLFGTFLCNSEQQRGKENLPKRTVSLWSYINSQLEDFTNPLYGSYSNHVLYPVASMRHLELWVGYYIRWNPRMKPQEPIHNRYKELLAKRAELQKKVEELQREISNRSTSSSERASSPAQCVTPVQTVVSpezifikation
NP_958435.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMT4B/CMT4B1/KIAA1073 | |
MTMR2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8898 | |
MTMR2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEEPPLLPGENIKDMAKDVTYICPFTGAVRGTLTVTNYRLYFKSMERDPPFVLDASLGVINRVEKIGGASSRGENSYGLETVCKDIRNLRFAHKPEGRTRRSIFENLMKYAFPVSNNLPLFAFEYKEVFPENGWKLYDPLLEYRRQGIPNESWRITKINERYELCDTYPALLVVPANIPDEELKRVASFRSRGRIPVLSWIHPESQATITRCSQPMVGVSGKRSKEDEKYLQAIMDSNAQSHKIFIFDARPSVNAVANKAKGGGYESEDAYQNAELVFLDIHNIHVMRESLRKLKEIVYPNIEETHWLSNLESTHWLEHIKLILAGALRIADKVESGKTSVVVHCSDGWDRTAQLTSLAMLMLDGYYRTIRGFEVLVEKEWLSFGHRFQLRVGHGDKNHADADRSPVFLQFIDCVWQMTRQFPTAFEFNEYFLITILDHLYSCLFGTFLCNSEQQRGKENLPKRTVSLWSYINSQLEDFTNPLYGSYSNHVLYPVASMRHLELWVGYYIRWNPRMKPQEPIHNRYKELLAKRAELQKKVEELQREISNRSTSSSERASSPAQCVTPVQTVV | |
RUO | |
MTMR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |