missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MUSK Partial ORF (NP_005583, 301 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00004593-Q01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16181485

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Intercellular communication is often mediated by receptors on the surface of one cell that recognize and are activated by specific protein ligands released by other cells. Members of one class of cell surface receptors, receptor tyrosine kinases (RTKs), are characterized by having a cytoplasmic domain containing intrinsic tyrosine kinase activity. This kinase activity is regulated by the binding of a cognate ligand to the extracellular portion of the receptor. DeChiara et al. (1996) [PubMed 8653786] noted that the RTKs, known to be expressed in cell type-specific fashions, play a role critical for the growth and differentiation of those cell types. For example, members of the neural-specific TRK family that recognize nerve growth factor are absolutely required for the survival and development of discrete neuronal subpopulations, and the receptor tyrosine kinases TIE1 (MIM 600222) and TIE2 (MIM 600221) play a critical role in the development of normal blood vessels.[supplied by OMIM]

Sequence: ISIAEWSKPQKDNKGYCAQYRGEVCNAVLAKDALVFLNTSYADPEEAQELLVHTAWNELKVVSPVCRPAAEALLCNHIFQECSPGVVPTPIPICREYCLA
Spezifikation

Spezifikation

NP_005583
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ISIAEWSKPQKDNKGYCAQYRGEVCNAVLAKDALVFLNTSYADPEEAQELLVHTAWNELKVVSPVCRPAAEALLCNHIFQECSPGVVPTPIPICREYCLA
RUO
MUSK
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4593
MUSK (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126323/MGC126324
MUSK
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote