Learn More
Human MVB12A (FAM125A) knockout HeLa cell lysate, 1 Kit
Product Code. 32157617 Shop All Abcam Products
Product Code. 32157617 Supplier Abcam Supplier No. ab2632671Kit
 Restricted Item
Verfügbar ab: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human MVB12A (FAM125A) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: MVB12A (FAM125A) knockout HeLa