Learn More
Abnova™ Human MX2 Partial ORF (NP_002454.1, 521 a.a. - 620 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004600-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene has a nuclear and a cytoplasmic form and is a member of both the dynamin family and the family of large GTPases. The nuclear form is localized in a granular pattern in the heterochromatin region beneath the nuclear envelope. A nuclear localization signal (NLS) is present at the amino terminal end of the nuclear form but is lacking in the cytoplasmic form due to use of an alternate translation start codon. This protein is upregulated by interferon-alpha but does not contain the antiviral activity of a similar myxovirus resistance protein 1. [provided by RefSeq]
Sequence: QKAMEIIQQAFINVAKKHFGEFFNLNQTVQSTIEDIKVKHTAKAENMIQLQFRMEQMVFCQDQIYSVVLKKVREEIFNPLGTPSQNMKLNSHFPSNESSVSpezifikation
NP_002454.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QKAMEIIQQAFINVAKKHFGEFFNLNQTVQSTIEDIKVKHTAKAENMIQLQFRMEQMVFCQDQIYSVVLKKVREEIFNPLGTPSQNMKLNSHFPSNESSV | |
RUO | |
MX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4600 | |
MX2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MXB | |
MX2 | |
Recombinant | |
wheat germ expression system |