missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human N6AMT2 (aa 72-144) Control Fragment Recombinant Protein

Artikelnummer. 30181171
Change view
Click to view available options
Menge:
100 μl
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30181171 100 μl 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30181171 Lieferant Invitrogen™ Lieferanten-Nr. RP97984

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60017 (PA5-60017. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, 3 families of mammalian DNA methyltransferase genes have been identified which include Dnmt1, Dnmt2 and Dnmt3. Dnmt1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. Dnmt2 is expressed at low levels in adult tissues and its inactivation does not affect DNA methylation or maintenance of methylation. Dnmt2 contains all the sequence motifs diagnostic of DNA (cytosine-5)-methyltransferases but appears to lack the large N-terminal regulatory domain common to other eukaryotic methyltransferases.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q8WVE0
Konzentration ≥5.0 mg/mL
Zur Verwendung mit (Anwendung) Blocking Assay, Control
Zusammensetzung 1 M urea, PBS with no preservative; pH 7.4
Gen-ID (Entrez) 221143
Name Human N6AMT2 (aa 72-144) Control Fragment
Menge 100 μl
Kennzeichnung RUO
Gen-Alias 2510005D08Rik; AW045965; Ayu21-96; DMNT2; DNMT2; EEF1A lysine methyltransferase 1; Eef1akmt1; ESP13; eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1; Gt(Ayu21)96 Imeg; Gt(pU21)96 Imeg; GtAyu21-96; M.HsaIIP; N(6)-adenine-specific DNA methyltransferase 2; N-6 adenine-specific DNA methyltransferase 2 (putative); N6AMT2; Protein-lysine N-methyltransferase N6AMT2; PuMet; RGD1306762; RNMT1; RP11-406H21.1
Gebräuchliche Bezeichnung N6AMT2
Gensymbol Eef1akmt1
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung His-ABP-tag
Sequenz GEGGRIACVSAPSVYQKLRELCRENFSIYIFEYDKRFAMYGEEFIFYDYNNPLDLPERIAAHSFDIVIADPPY
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Expressionssystem E. coli
Form Liquid
Reinheits- oder Qualitätsgrad >80% by SDS-PAGE and Coomassie blue staining
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.