Learn More
Abnova™ Human NCOA3 Partial ORF (NP_006525, 251 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008202-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a nuclear receptor coactivator that interacts with nuclear hormone receptors to enhance their transcriptional activator functions. The encoded protein has histone acetyltransferase activity and recruits p300/CBP-associated factor and CREB binding protein as part of a multisubunit coactivation complex. This protein is initially found in the cytoplasm but is translocated into the nucleus upon phosphorylation. Two transcript variants encoding different isoforms have been found for this gene. In addition, a polymorphic repeat region is found in the C-terminus of the encoded protein. [provided by RefSeq]
Sequence: RRITTGERTFPSNPESFITRHDLSGKVVNIDTNSLRSSMRPGFEDIIRRCIQRFFSLNDGQSWSQKRHYQEAYLNGHAETPVYRFSLADGTIVTAQTKSKLFRNPVTNDRSpezifikation
NP_006525 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RRITTGERTFPSNPESFITRHDLSGKVVNIDTNSLRSSMRPGFEDIIRRCIQRFFSLNDGQSWSQKRHYQEAYLNGHAETPVYRFSLADGTIVTAQTKSKLFRNPVTNDR | |
RUO | |
NCOA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8202 | |
NCOA3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACTR/AIB-1/AIB1/CAGH16/CTG26/KAT13B/MGC141848/RAC3/SRC3/TNRC14/TNRC16/TRAM-1/bHLHe42/pCIP | |
NCOA3 | |
Recombinant | |
wheat germ expression system |