Learn More
ACROBiosystems Human NKp30 / NCR3 / CD337 Protein, Fc Tag
338.70 € - 2286.25 €
Spezifikation
Zugriffsnummer | AAH52582 |
---|---|
Molekulargewicht | 39.5 kDa |
Quelle | Human NKp30, Fc Tag (NC3-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Gly 135 (Accession # AAH52582). |
Immunogen | Leu 19 - Gly 135 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15887337
|
ACROBiosystems
NC3-H5259-100UG |
100 μg |
338.70 €
100 Mikrogramm |
Verfügbar ab: 03-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15897337
|
ACROBiosystems
NC3-H5259-1MG |
1 mg |
2286.25 €
1 Milligramm |
Verfügbar ab: 03-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Natural cytotoxicity triggering receptor 3 (NCR3) is also known as Activating natural killer receptor p30, Natural killer cell p30-related protein (NK-p30), CD antigen CD337, 1C7, LY117. NCR3 /CD337 /NKp30 belongs to the natural cytotoxicity receptor (NCR) family. NCR3 /NKp30 contains one Ig-like (immunoglobulin-like) domain. NCR3 /NKp30 is selectively expressed by all resting and activated NK cells and weakly expressed in spleen. NCR3 is homodimer in the unliganted form. NCR3 interacts with CD3Z. NCR3 interacts with and is activated by binding to NCR3LG1 or BAG6. Engagement of NCR3 by BAG6 also promotes dendritic cell (DC) maturation, both through killing those DCs that did not properly acquire a mature phenotype, and inducing NK cells to release TNFA and IFNG, which promotes DC maturation.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.5 kDa. The protein migrates as 50-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH52582 | |
Human NKp30, Fc Tag (NC3-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Gly 135 (Accession # AAH52582). | |
-20°C | |
NKp30 | |
Human | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
39.5 kDa | |
Leu 19 - Gly 135 | |
NCR3,CD337,NKp30,1C7,LY117,MALS | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |